Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Pyrazophos, MilliporeSigma™ Supelco™

Catalog No. 111019101 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9101 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9101 Supplier MilliporeSigma Supelco Supplier No. 45648250MG
Add to Cart
Add to Cart

Pyrazophos belongs to the class of organophosphorus pesticides, widely used in agriculture and household plant care, with a broad spectrum of action and shorter persistence in the environment compared to organochlorine pesticides.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 13457-18-6
Physical Form Neat
Linear Formula C14H20N3O5 PS
MDL Number MFCD00055494
Type Analytical Standard
Quantity 250 mg
Formula Weight 373.36
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.