Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Quinchlorac, MilliporeSigma™ Supelco™

Catalog No. 111015660 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5660 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5660 Supplier MilliporeSigma Supelco Supplier No. 36521250MG
Add to Cart
Add to Cart

Quinchlorac is a postemergence herbicide, used for the control of broadleaf weeds, especially white clover (Trifolium repens L.) and Veronica filiformis.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 84087-01-4
Flash Point 100°C
Physical Form Neat
Linear Formula C10H5 Cl2 NO2
MDL Number MFCD00072495
Synonym 3,7-Dichloro-8-quinolinecarboxylic acid
Type Analytical Standard
Quantity 250 mg
Formula Weight 242.06
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.