Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Quizalofop-ethyl, MilliporeSigma™ Supelco™

Catalog No. 111020305 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0305 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0305 Supplier MilliporeSigma Supelco Supplier No. 3430650MG
Add to Cart
Add to Cart

Quizalofop-ethyl is a phenoxy propionate herbicide, which is used to control both annual and perennial grass weeds in broadleaved crops.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 76578-14-8
Flash Point 100°C
Physical Form Neat
Linear Formula C19H17 ClN2O4
MDL Number MFCD00128062
Synonym Ethyl 2-[4-(6-chloro-2-quinoxalinyloxy)phenoxy]propionate
Type Analytical Standard
Quantity 50 mg
Formula Weight 372.80
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.