Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Resmethrin, Mixture of isomers, MilliporeSigma™ Supelco™

Catalog No. 111019382 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9382 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9382 Supplier MilliporeSigma Supelco Supplier No. 45655250MG
Only null left
Add to Cart
Add to Cart

Resmethrin is a pyrethroid insecticide, used as a tool for the pest management.

Agriculture and Environmental
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 10453-86-8
Physical Form Neat
Linear Formula C22H26O3
MDL Number MFCD00041806
Type Analytical Standard
Quantity 250 mg
Formula Weight 338.44
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.