Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Simazine Solution, 100 μg/mL in Methanol, MilliporeSigma™ Supelco™

Catalog No. 111020007 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0007 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0007 Supplier MilliporeSigma Supelco Supplier No. 365882ML
Add to Cart
Add to Cart

Simazine is a systemic triazine herbicide, which can have the tendency to suppress the photosynthesis process and block the linear electron transport in light chain.

Agriculture, Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 122-34-9
Linear Formula C7H12 ClN5
MDL Number MFCD00023174
Type Analytical Standard
Quantity 2 mL
Formula Weight 201.66
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 100 μg/mL in methanol

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.