Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Spirotetramat Metabolite BYI08330-cis-enol, MilliporeSigma™ Supelco™

Catalog No. 111018067 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8067 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8067 Supplier MilliporeSigma Supelco Supplier No. 3248410MG
Add to Cart
Add to Cart

Spirotetramat-enol [cis-3-(2,5-dimethylphenyl)-4-hydroxy-8-methoxy-1-azaspiro[4,5]dec-3-en-2-one] is the major metabolite of spirotetramat in plants.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 203312-38-3
Physical Form Neat
Linear Formula C18H23 NO3
Synonym cis-3-(2,5-Dimethylphenyl)-4-hydroxy-8-methoxy-1-azaspiro[4.5 ]dec-3-en-2-one
Type Analytical Standard
Quantity 10 mg
Formula Weight 301.38
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.