Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Spirotetramat Metabolite BYI08330-mono-hydroxy, MilliporeSigma™ Supelco™

Catalog No. 111018085 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8085 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8085 Supplier MilliporeSigma Supelco Supplier No. 3248610MG
Add to Cart
Add to Cart

Spirotetramat is the member of cyclic ketoenoles. It is an inhibitor of acetyl-CoA carboxylase which is active against a wide spectrum of sucking insects.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 1172134-12-1
Physical Form Neat
Linear Formula C18H25 NO3
Synonym cis-3-(2,5-Dimethylphenyl)-4-hydroxy-8-methoxy-1-azaspiro[4.5 ]decan-2-one
Type Analytical Standard
Quantity 10 mg
Formula Weight 303.40
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.