Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Spiroxamine, MilliporeSigma™ Supelco™

Catalog No. 111019350 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9350 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9350 Supplier MilliporeSigma Supelco Supplier No. 46443100MG
Add to Cart
Add to Cart

Spiroxamine belongs to the class of spiroketalamines. It is widely used as a broad-spectrum fungicide in cereals and grapes with selective, protective, curative and eradicative properties against powdery mildew.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 118134-30-8
Flash Point 147°C
Physical Form Neat
Linear Formula C18H35 NO2
MDL Number MFCD03095708
Synonym N-Ethyl-N-propyl-8-tert-butyl-1,4-dioxaspiro[4.5 ]dec-2-ylmethylamine
Type Analytical Standard
Quantity 100 mg
Formula Weight 297.48
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.