Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Sulfaquinoxaline, MilliporeSigma™ Supelco™

Catalog No. 111014798 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4798 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4798 Supplier MilliporeSigma Supelco Supplier No. 45662250MG
Add to Cart
Add to Cart

Sulfaquinoxaline belongs to the class of sulfonamides. It is a feed sample additive, which is used for the prevention or treatment of poultry leucocytozoonosis and coccidiosis.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 59-40-5
Physical Form Neat
Linear Formula C14H12N4O2 S
MDL Number MFCD00055406
Synonym 4-Amino-N-(2-quinoxalinyl)benzenesulfonamide; Sulfaquinoxalin
Type Analytical Standard
Quantity 250 mg
Formula Weight 300.34
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.