Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Sulfur, MilliporeSigma™ Supelco™

Catalog No. 111020542 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0542 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0542 Supplier MilliporeSigma Supelco Supplier No. 36576250MG
Add to Cart
Add to Cart

Sulfur can play the role as a cathode material and can also find applications in electric and hybrid electric vehicles. It can also form a composite with graphite, which can serve as an improved sulfur cathode materials for lithium-sulfur(Li-S) batteries with higher capacity and cycling stability.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
Ignition Point 450°F
CAS 7704-34-9
Vapor Pressure 1 mmHg (183.8°C)
Vapor Density 8.9 (vs. air)
Flash Point 160°C
Physical Form Neat
Linear Formula S
MDL Number MFCD00085316
Melting Point 112.8°C (rhombic); 117°C to 120°C; 119.0°C (monoclinic)
Boiling Point 444.7°C
Type Analytical Standard
Quantity 250 mg
Formula Weight 32.07
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.