Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Tebuconazole-(tert-butyl-d9), ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111018078 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8078 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8078 Supplier MilliporeSigma Supelco Supplier No. 556395MG
Add to Cart
Add to Cart
Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 1246818-83-6
Linear Formula C16D9H13 ClN3 O
MDL Number MFCD28137842
Synonym Tebuconazole-d9
Type Analytical Standard
Quantity 5 mg
Formula Weight 316.87
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.