Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Tebutam, MilliporeSigma™ Supelco™

Catalog No. 111019566 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9566 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9566 Supplier MilliporeSigma Supelco Supplier No. 36566250MG
Add to Cart
Add to Cart

Tebutam belongs to the class of acetamide herbicides and is commonly applied to the soil surface, for the control of weeds in rape farming.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 35256-85-0
Flash Point 80°C - Closed Cup
Physical Form Neat
Linear Formula C15H23 NO
MDL Number MFCD00145351
Synonym N-Benzyl-N-isopropylpivalamide
Type Analytical Standard
Quantity 250 mg
Formula Weight 233.35
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.