Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Terbufos-Sulfone Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022453 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2453 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2453 Supplier MilliporeSigma Supelco Supplier No. 31580100MG

Terbufos-sulfone is one of the major metabolites of organophosphorus pesticide terbufos. It is found to be toxic for living beings, even at very low levels.

TRUSTED_SUSTAINABILITY

Specifications

CAS 56070-16-7
Flash Point Not Applicable
Molecular Formula C9H21O4PS3
Linear Formula C9H21O4PS3
MDL Number MFCD01863139
Quantity 100 mg
UN Number UN 2810 6.1 / PGI
Formula Weight 320.43
Grade Analytical
Physical Form Neat

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.