Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Tetraconazole, MilliporeSigma™ Supelco™

Catalog No. 111018331 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8331 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8331 Supplier MilliporeSigma Supelco Supplier No. 37087100MG
Add to Cart
Add to Cart
Agriculture, Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 112281-77-3
Physical Form Neat
Linear Formula C13H11 Cl2F4N3 O
MDL Number MFCD01632314
Synonym (RS)-2-(2,4-Dichlorophenyl)-3-(1 H-1,2,4-triazolyl)propyl 1,1,2,2-tetrafluoroethyl ether; (+/-)-1-[2-(2,4-Dichlorophenyl)-3-(1,1,2,2-tetrafluoroethoxy)propyl]-1 H-1,2,4-triazole
Type Analytical Standard
Quantity 100 mg
Formula Weight 372.15
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.