Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Thiacloprid-(thiazolidin ring-d4), ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002493 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-2493 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-2493 Supplier MilliporeSigma Supelco Supplier No. 3067310MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Find more information here: Neonicotinoids

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 1793071-39-2
Linear Formula C10D4H5 ClN4 S
Synonym Thiacloprid-d4; [3-(6-Chloro-3-pyridinylmethyl)-2-thiazolidin-d4-ylidene]cyanamide
Type Analytical Standard
Quantity 10 mg
Formula Weight 256.75
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.