Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Thiamethoxam-d3, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111018388 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8388 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8388 Supplier MilliporeSigma Supelco Supplier No. 3817625MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Find more information here: Neonicotinoids

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
Product Line PESTANAL
CAS 1294048-82-0
Physical Form Neat
Linear Formula C8D3H7 ClN5O3 S
MDL Number MFCD22565806
Synonym 3-(2-Chloro-5-thiazolylmethyl)tetrahydro-5-methyl-d3-N-nitro-4 H-1,3,5-oxadiazin-4-imine
Type Analytical Standard
Quantity 25 mg
Formula Weight 294.73
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.