Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Thiencarbazone-methyl, MilliporeSigma™ Supelco™

Catalog No. 111018651 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8651 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8651 Supplier MilliporeSigma Supelco Supplier No. 32721100MG
Add to Cart
Add to Cart

Thiencarbazone-methyl is a pesticide, which is used for controlling weeds in corn plantation.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 317815-83-1
Physical Form Neat
Linear Formula C12H14N4O7S2
MDL Number MFCD15146774
Synonym Methyl 4-[(4,5-dihydro-3-methoxy-4-methyl-5-oxo-1 H-1,2,4-triazol-1-yl)carbonylsulfamoyl]-5-methylthiophene-3-carboxylate
Type Analytical Standard
Quantity 100 mg
Formula Weight 390.39
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.