Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Tolfenpyrad, MilliporeSigma™ Supelco™

Catalog No. 111018726 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8726 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8726 Supplier MilliporeSigma Supelco Supplier No. 3704325MG
Add to Cart
Add to Cart

Tolfenpyrad is an effective insecticide, used against pests that are resistant to existing insecticides such as organophosphates and carbamates.

TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 129558-76-5
Physical Form Neat
Linear Formula C21H22 ClN3O2
MDL Number MFCD08273850
Synonym 4-Chloro-3-ethyl-1-methyl-N-[4-(p-tolyloxy)benzyl]pyrazole-5-carboxamide; 4-Chloro-3-ethyl-1-methyl-N-{[4-(4-methylphenoxy)phenyl]methyl}-1 H-pyrazole-5-carboxamide
Type Analytical Standard
Quantity 25 mg
Formula Weight 383.87
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.