Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Triclosan-methyl, MilliporeSigma™ Supelco™

Catalog No. 111018442 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8442 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8442 Supplier MilliporeSigma Supelco Supplier No. 3422850MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Analytical standard used as a positive control in antimicrobial assays.

Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Triclosan-methyl
CAS 1-1-4640
Product Line PESTANAL
Physical Form Neat
Linear Formula C13H9 Cl3O2
Technique Gas Chromatography
MDL Number MFCD04039700
Synonym 2,4,4 ′-Trichloro-2 ′-methoxydiphenyl ether; 5-Chloro-2-(2,4-dichlorophenoxy)anisole; Methyl triclosan
Type Analytical Standard
Industry Type Environmental Testing
Molecular Weight (g/mol) 303.57
Quantity 50 mg
Formula Weight 303.57
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.