Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Triethyl phosphate, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111015543 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5543 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5543 Supplier MilliporeSigma Supelco Supplier No. 194451ML
Environmental
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
Ignition Point 845°F
CAS 78-40-0
Vapor Pressure 1 mmHg (40°C)
Vapor Density 6.28 (vs. air)
Density 1.072 g/mL (at 25°C)
Linear Formula (C2H5 O)3 PO
MDL Number MFCD00009077
Boiling Point 215°C
Synonym Ethyl phosphate
Type Analytical Standard
Quantity 1 mL
Formula Weight 182.15
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.