Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Valifenalate, Mixture of isomers, MilliporeSigma™ Supelco™

Catalog No. 111013816 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3816 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3816 Supplier MilliporeSigma Supelco Supplier No. 3707825MG
Add to Cart
Add to Cart

Valifenalate is a fungicide, which can be used to control downy mildew disease caused by Plasmopara viticola fungi in vines.

Agriculture
TRUSTED_SUSTAINABILITY

Specifications

Product Line PESTANAL
CAS 283159-90-0
Physical Form Neat
Linear Formula C19H27 ClN2O5
MDL Number MFCD22373630
Synonym N-[(1-Methylethoxy)carbonyl]-L-valyl-3-(4-chlorophenyl)-beta-alanine methyl ester; Methyl N-(isopropoxycarbonyl)-L-valyl-(3 RS)-3-(4-chlorophenyl)-beta-alaninate
Type Analytical Standard
Quantity 25 mg
Formula Weight 398.88
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.