Learn More
Phenomenex Inc PestiCarb/NH2 SPE Column
Catalog No. 502400672 Shop All Phenomenex Products
Encompass_Preferred
PestiCarb/NH2 SPE Column 500mg/500mg/6mL, 100 m2/g PC
Catalog No. 50-240-0672 Supplier Phenomenex Inc Supplier No. PN000610
PestiCarb/NH2 SPE Column 500mg/500mg/6mL, 100 m2/g PC