Learn More
AccuStandard, Inc PESTICIDE A MIX 1ML
SDP
Catalog No. NC9461441
Encompass
Single-Column Analytes Mix. 1 mL. 0.2 mg/mL in MeOH.
Catalog No. NC9461441 Supplier AccuStandard, Inc Supplier No. M8081SC
Single-Column Analytes Mix. 1 mL. 0.2 mg/mL in MeOH.