Learn More
Thomas Scientific Pestle Motor Mixer, Battery-Powered, with Grooved Grip Handle
SDP
Catalog No. NC1002749
Encompass
The Argos Pellet Mixer is a lightweight battery powered instrument that can be used with disposable pestles, mixers and micro tubes. Cell tissue and pellets are completely homogenized within seconds by means of a high speed vortexing action. The pestle features a grooved grip handle tailored to fit the microtube. The grooved grip makes the handle easy to turn without slipping, even when wearing gloves. Greatly reduces hand fatigue DNase-, RNase-, and pyrogen-free Pestle length: 7.5cm (3") Comes complete with 2 "AA"batteries and Pestle/Mixer adapter
Catalog No. NC1002749 Supplier Thomas Scientific Supplier No. 1226W45
The Argos Pellet Mixer is a lightweight battery powered instrument that can be used with disposable pestles, mixers and micro tubes. Cell tissue and pellets are completely homogenized within seconds by means of a high speed vortexing action. The pestle features a grooved grip handle tailored to fit the microtube. The grooved grip makes the handle easy to turn without slipping, even when wearing gloves. Greatly reduces hand fatigue DNase-, RNase-, and pyrogen-free Pestle length: 7.5cm (3") Comes complete with 2 "AA"batteries and Pestle/Mixer adapter