Learn More
Electron Microscopy Sciences Potter-Elvehjem Tissue Grinder Pestle Serrated, 55 ml, 30 mmx 130 mm Mortor, 266 mm OAL
SDP
Catalog No. 501900333
Encompass_Preferred
Replacement part for Potter-Elvehjem Tissue Grinder with radial serrations.
Size: 55 ml
Mortor: 30 mmx 130 mm
OAL: 266 mm
Catalog No. 50-190-0333 Supplier Electron Microscopy Sciences Supplier No. 6479585
Replacement part for Potter-Elvehjem Tissue Grinder with radial serrations.
Size: 55 ml
Mortor: 30 mmx 130 mm
OAL: 266 mm