Learn More
Chemglass Life Sciences Pestle with 1.5 mL microcentrifuge tube, polypropylene, pack of 100
SDP
Catalog No. NC3669322
Encompass_Preferred
This disposable polypropylene pestle is paired with a 1.5 mL microcentrifuge tube and supplied in packs of 100. The matched pestle and tube are designed to improve resuspension of protein and DNA pellets and to aid soft tissue grinding; pestles are suitable for manual use or with motorized mixers.
  • Disposable polypropylene construction for low contamination risk.
  • DNase, RNase, and pyrogen-free to maintain sample integrity.
  • Autoclavable for optional sterilization prior to use.
  • Pestle shafts suitable for motorized mixer use or manual operation.
  • Microtubes with positive snap-cap seal and molded graduations for accurate volume reading.
  • Frosted sidewall marking spots and flat caps pierceable by syringe for easy labeling and access.
  • Individually bagged and over-bagged; bulk packed with lot control and material traceability.
Catalog No. NC3669322 Supplier Chemglass Life Sciences Supplier No. CLS5002B35
This disposable polypropylene pestle is paired with a 1.5 mL microcentrifuge tube and supplied in packs of 100. The matched pestle and tube are designed to improve resuspension of protein and DNA pellets and to aid soft tissue grinding; pestles are suitable for manual use or with motorized mixers.
  • Disposable polypropylene construction for low contamination risk.
  • DNase, RNase, and pyrogen-free to maintain sample integrity.
  • Autoclavable for optional sterilization prior to use.
  • Pestle shafts suitable for motorized mixer use or manual operation.
  • Microtubes with positive snap-cap seal and molded graduations for accurate volume reading.
  • Frosted sidewall marking spots and flat caps pierceable by syringe for easy labeling and access.
  • Individually bagged and over-bagged; bulk packed with lot control and material traceability.