Learn More
Agilent Technologies PET-11 A-D VECTORS
Supplier:  Agilent Technologies 211523
Encompass
pET-11 a-d Vectors. Tightly controlled T7 RNA polymerase-based gene expression in E. coli. Convenient cloning sites for optimal expression of cloned protein-coding sequence. BL21(DE3) and BL21(DE3)pLysS competent cells are provided.
Catalog No. 50-125-242
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.