Learn More
Applied Biophysics Inc 96W20IDF PET ARRAYS 6/PK
SDP
Catalog No. NC0758945
Encompass
  • 96 well array format.
  • Inter-digitated finger (idf) electrode configuration.
  • Polyethylene terephthalate (PET) substrate material.
  • Substrate thickness of 0.25 mm.
  • Total electrode area per well of 3.985 mm².
  • Measures 4,000-8,000 cells per well.
  • Well top assembly made of polystyrene.
  • Applications include cell-ECM protein interactions and signal transduction assays.
  • Includes 6 arrays per package.
Catalog No. NC0758945 Supplier Applied Biophysics Inc Supplier No. 96W20IDF PET
  • 96 well array format.
  • Inter-digitated finger (idf) electrode configuration.
  • Polyethylene terephthalate (PET) substrate material.
  • Substrate thickness of 0.25 mm.
  • Total electrode area per well of 3.985 mm².
  • Measures 4,000-8,000 cells per well.
  • Well top assembly made of polystyrene.
  • Applications include cell-ECM protein interactions and signal transduction assays.
  • Includes 6 arrays per package.