Learn More
ATHENA ES PET COLD TRYPSIN CELL RELEASE
Catalog No. NC1017857
Encompass_Preferred
​PET™ is a superior cell dissociation reagent specially formulated for use under “Cold Trypsin” conditions. The unique blend of polyvinylpyrrolidone, EGTA, and trypsin in a HBS base allows the trypsin to retain enzymatic activity under cool temperatures, unlike standard trypsin which requires a temperature of 37°C. When used to detach monolayer cultures of epithelial cells from the growth substratum, this reagent yields more viable cells from monolayers grown in serum-free medium than traditional trypsin formulas.
Catalog No. NC1017857 Supplier ATHENA ES Supplier No. 0405
​PET™ is a superior cell dissociation reagent specially formulated for use under “Cold Trypsin” conditions. The unique blend of polyvinylpyrrolidone, EGTA, and trypsin in a HBS base allows the trypsin to retain enzymatic activity under cool temperatures, unlike standard trypsin which requires a temperature of 37°C. When used to detach monolayer cultures of epithelial cells from the growth substratum, this reagent yields more viable cells from monolayers grown in serum-free medium than traditional trypsin formulas.