Learn More
Research Products International Corp PET Media Square Bottles, Sterile, 125 ml, 48 per Case
SDP
Catalog No. 50998617
Encompass
These square PET media bottles offer the clarity of glass with the durability of plastic and a space saving design. Improved features include a wide mouth opening, thicker wall dimensions and a leak proof double seal closure. Bottles have permanent molded in graduations and dripless pour lip. Bottles and caps are pre-sterilized and tested for pH stability, temperature durability, cloning efficiency and cytotoxicity. Supplied with caps attached in shrink-wrapped trays.
Catalog No. 50-998-617 Supplier Research Products International Corp Supplier No. 219975
These square PET media bottles offer the clarity of glass with the durability of plastic and a space saving design. Improved features include a wide mouth opening, thicker wall dimensions and a leak proof double seal closure. Bottles have permanent molded in graduations and dripless pour lip. Bottles and caps are pre-sterilized and tested for pH stability, temperature durability, cloning efficiency and cytotoxicity. Supplied with caps attached in shrink-wrapped trays.