Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ PET Round Bottle

Catalog No. 02993999
Change view
Click to view available options
Capacity (Metric):
480 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
02-993-999 480 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-993-999 Supplier Qorpak™ Supplier No. PLC06988
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ideal for oil analysis. Qorpak™ PET Round Bottle makes content identification easy.

  • Provide good alcohol and solvent barrier
  • PET is stress and impact resistant

Specifications

Capacity (Metric) 480 mL
Color Clear
Material PET
Sterility Non-sterile
Closure Color White
Neck Finish 70-400
Closure Included Yes
Graduated No
Mouth Wide, Threaded
Closure Liner Polyethylene Foam
Closure Material Polypropylene
Autoclavable Not Autoclavable
Capacity (English) 16 oz.
Shape Round
Product Type Packer Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.