Learn More
Medchemexpress LLC HY-103349 10mg Medchemexpress, PETCM CAS:10129-56-3 Purity:>98%
SDP
Catalog No. 501968077
Encompass_Preferred
Medchemexpress, HY-103349 10mg PETCM CAS:10129-56-3 PETCM is an activator of caspase-3. PETCM can induces cell apoptosis and stimulate apoptosome formation in HeLa cell cytosols. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8077 Supplier Medchemexpress LLC Supplier No. HY10334910MG
Add to Cart
Add to Cart
Medchemexpress, HY-103349 10mg PETCM CAS:10129-56-3 PETCM is an activator of caspase-3. PETCM can induces cell apoptosis and stimulate apoptosome formation in HeLa cell cytosols. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.