Learn More
Sterlitech Corporation Polyester (PETE) Drain Disc, 142mm, 25/Pk
SDP
Catalog No. 501952367
Encompass_Preferred
Polyester Drain Discs increase flow rate and throughputs by preventing blockage or "pore blinding" of the capillary pores in screen membranes.
Catalog No. 50-195-2367 Supplier Sterlitech Corporation Supplier No. PETEDD14225
Polyester Drain Discs increase flow rate and throughputs by preventing blockage or "pore blinding" of the capillary pores in screen membranes.