Learn More
Sterlitech Corporation Polyester (PETE) Drain Disc, 293mm, 25/Pk
SDP
Catalog No. 501952368
Encompass_Preferred
These Polyester Drain Discs increase flow rate and throughputs by preventing blockage or "pore blinding" of the capillary pores in screen membranes.
Catalog No. 50-195-2368 Supplier Sterlitech Corporation Supplier No. PETEDD29325
These Polyester Drain Discs increase flow rate and throughputs by preventing blockage or "pore blinding" of the capillary pores in screen membranes.