Learn More
Sterlitech Corporation Polyester (PETE) Drain Disc (Mesh Spacer), 47mm, 100/Pk
SDP
Catalog No. 501952366
Encompass_Preferred
These Polyester Drain Discs increase flow rate and throughputs by preventing blockage or "pore blinding" of the capillary pores in screen membranes.
Catalog No. 50-195-2366 Supplier Sterlitech Corporation Supplier No. PETEDD47100
Add to Cart
Add to Cart
These Polyester Drain Discs increase flow rate and throughputs by preventing blockage or "pore blinding" of the capillary pores in screen membranes.