Learn More
Sterlitech Corporation Polyester (PETE) Drain Disc, 90mm, 25/Pk
SDP
Catalog No. 501952361
Encompass_Preferred
These Polyester Drain Discs increase flow rate and throughputs by preventing blockage or "pore blinding" of the capillary pores in screen membranes.
Catalog No. 50-195-2361 Supplier Sterlitech Corporation Supplier No. PETEDD9025
These Polyester Drain Discs increase flow rate and throughputs by preventing blockage or "pore blinding" of the capillary pores in screen membranes.