Learn More
Sterlitech Corporation Polyester (PETE) Membrane Filters, 0.4 Micron, 47mm, 100/Pk
SDP
Catalog No. 501952365
Encompass_Preferred
Sterlitech polyester track etch (PETE) membrane filters, 0.4 micron pore size, 47 mm diameter, pack of 100. Hydrophilic, solvent resisant film for blood assays, microscopy, and highly caustic samples.
Catalog No. 50-195-2365 Supplier Sterlitech Corporation Supplier No. PET0447100
Sterlitech polyester track etch (PETE) membrane filters, 0.4 micron pore size, 47 mm diameter, pack of 100. Hydrophilic, solvent resisant film for blood assays, microscopy, and highly caustic samples.