Learn More
Sterlitech Corporation Polyester (PETE) Membrane Filters, 10.0 Micron, 47mm, 100/Pk
SDP
Catalog No. 501952364
Encompass_Preferred
Sterlitech polyester track etch (PETE) membrane filters, 10 micron pore size, 47 mm diameter, pack of 100. Hydrophilic, solvent resisant film for blood assays, microscopy, and highly caustic samples.
Catalog No. 50-195-2364 Supplier Sterlitech Corporation Supplier No. PET10047100
Sterlitech polyester track etch (PETE) membrane filters, 10 micron pore size, 47 mm diameter, pack of 100. Hydrophilic, solvent resisant film for blood assays, microscopy, and highly caustic samples.