Learn More
Sterlitech Corporation Polyester (PETE) Membrane Filters, 10.0 Micron, 90mm, 30/Pk
SDP
Catalog No. 501952363
Encompass_Preferred
Sterlitech polyester track etch (PETE) membrane filters, 10 micron pore size, 90 mm diameter, pack of 30. Hydrophilic, solvent resisant film for blood assays, microscopy, and highly caustic samples.
Catalog No. 50-195-2363 Supplier Sterlitech Corporation Supplier No. PET1009030
Sterlitech polyester track etch (PETE) membrane filters, 10 micron pore size, 90 mm diameter, pack of 30. Hydrophilic, solvent resisant film for blood assays, microscopy, and highly caustic samples.