Learn More
Sterlitech Corporation Polyester (PETE) Membrane Filters, 5.0 Micron, 25mm, 100/Pk
SDP
Catalog No. 501952360
Encompass_Preferred
Sterlitech polyester track etch (PETE) membrane filters, 5 micron pore size, 25 mm diameter, pack of 100. Hydrophilic, solvent resisant film for blood assays, microscopy, and highly caustic samples.
Catalog No. 50-195-2360 Supplier Sterlitech Corporation Supplier No. PET5025100
Sterlitech polyester track etch (PETE) membrane filters, 5 micron pore size, 25 mm diameter, pack of 100. Hydrophilic, solvent resisant film for blood assays, microscopy, and highly caustic samples.