Learn More
Sterlitech Corporation Polyester (PETE) Membrane Filters, 8.0 Micron, 47mm, 100/Pk
SDP
Catalog No. 501952362
Encompass_Preferred
Sterlitech polyester track etch (PETE) membrane filters, 8 micron pore size, 47 mm diameter, pack of 100. Hydrophilic, solvent resisant film for blood assays, microscopy, and highly caustic samples.
Catalog No. 50-195-2362 Supplier Sterlitech Corporation Supplier No. PET8047100
Add to Cart
Add to Cart
Sterlitech polyester track etch (PETE) membrane filters, 8 micron pore size, 47 mm diameter, pack of 100. Hydrophilic, solvent resisant film for blood assays, microscopy, and highly caustic samples.