Learn More
Sterlitech Corporation Polyester (PETE) Membrane Filters, 10.0 Micron, 25mm, 100/Pk
SDP
Catalog No. NC1694594
Encompass_Preferred
Sterlitech polyester track etch (PETE) membrane filters, 10 micron pore size, 25 mm diameter, pack of 100. Hydrophilic, solvent resisant film for blood assays, microscopy, and highly caustic samples.
Catalog No. NC1694594 Supplier Sterlitech Corporation Supplier No. PET10025100
Sterlitech polyester track etch (PETE) membrane filters, 10 micron pore size, 25 mm diameter, pack of 100. Hydrophilic, solvent resisant film for blood assays, microscopy, and highly caustic samples.