Learn More
Sterlitech Corporation Polyester (PETE) Membrane Filters, Hydrophobic, Nonwoven Support, 0.2 Micron, 13mm, 100/Pk
SDP
Catalog No. 501952359
Encompass_Preferred
Hydrophobic PETE membrane filters are a great option for venting applications where PTFE or polyolefin membrane filters have traditionally been utilized.Applications:* Vent filters for electrical enclosures* Vent filters for microbial retention in medical devices* Air/gas filtration while repelling liquid water* Air/gas filtration while repelling oils and solvents with surface tension >25 dynes/cm
Catalog No. 50-195-2359 Supplier Sterlitech Corporation Supplier No. 1300012
Hydrophobic PETE membrane filters are a great option for venting applications where PTFE or polyolefin membrane filters have traditionally been utilized.Applications:* Vent filters for electrical enclosures* Vent filters for microbial retention in medical devices* Air/gas filtration while repelling liquid water* Air/gas filtration while repelling oils and solvents with surface tension >25 dynes/cm