Learn More
Foxx Life Sciences EZBio Single Use Media Bottle Assembly, 60mL Sterilized PETG Bottles for Aseptic Sampling and Sterile Storage, 38-430 VersaCap with Vent and Dip Tube, 10/CS
GREENER_CHOICE
Catalog No. 502236446
Encompass_Preferred
EZBio Single Use Media Bottle Assembly, 60mL Sterilized PETG Bottles for Aseptic Sampling and Sterile Storage, 38-430 VersaCap with Vent and Dip Tube, 10/CS
This item is not returnable. View return policy
Catalog No. 50-223-6446 Supplier Foxx Life Sciences Supplier No. 4370002FLS
EZBio Single Use Media Bottle Assembly, 60mL Sterilized PETG Bottles for Aseptic Sampling and Sterile Storage, 38-430 VersaCap with Vent and Dip Tube, 10/CS