Learn More
Foxx Life Sciences EZBio 2L (2,000mL) PETG Media Bottles, Non-Sterile, Square Storage Bottles with 53mm (53B) Neck, Without Caps, 6/PK
GREENER_CHOICE
Catalog No. 501613233
Encompass_Preferred
EZBio PETG Non-Sterile Media Bottles are perfect for storage and shipment reagents and media. Bottles withstand temperatures down to -40C for freezing applications. Shatter-resistant and thick-walled. Assembled in a Class 10,000 Cleanroom. Packaged in double-bagged trays. 100% USP Class VI, BPA-free, ADCF-free materials. VersaCaps, including options with vented membranes or molded hose barbs, are available separately.
Catalog No. 50-161-3233 Supplier Foxx Life Sciences Supplier No. 1376513FLS
EZBio PETG Non-Sterile Media Bottles are perfect for storage and shipment reagents and media. Bottles withstand temperatures down to -40C for freezing applications. Shatter-resistant and thick-walled. Assembled in a Class 10,000 Cleanroom. Packaged in double-bagged trays. 100% USP Class VI, BPA-free, ADCF-free materials. VersaCaps, including options with vented membranes or molded hose barbs, are available separately.