Learn More
Foxx Life Sciences EZBio 1L (1,000mL) PETG Media Bottles, Sterile, Square Storage Bottles with 38-430mm White Closed VersaCap Closures, 12/PK
GREENER_CHOICE
Catalog No. 501613230
Encompass_Preferred
EZBio PETG Sterile Media Bottles are perfect for storage of sterile reagents and media, and can be used with EZBio Single-Use Cap Assemblies for aseptic transfer and sampling applications. Bottles withstand temperatures down to -40C for freezing applications. Gamma-irradiated. Assembled in a Class 10,000 Cleanroom. Packaged in double-bagged trays. 100% USP Class VI, BPA-free, ADCF-free materials. VersaCaps with vented membranes or molded hose barbs available separately.
Catalog No. 50-161-3230 Supplier Foxx Life Sciences Supplier No. 1375211FLS
EZBio PETG Sterile Media Bottles are perfect for storage of sterile reagents and media, and can be used with EZBio Single-Use Cap Assemblies for aseptic transfer and sampling applications. Bottles withstand temperatures down to -40C for freezing applications. Gamma-irradiated. Assembled in a Class 10,000 Cleanroom. Packaged in double-bagged trays. 100% USP Class VI, BPA-free, ADCF-free materials. VersaCaps with vented membranes or molded hose barbs available separately.