Learn More
Foxx Life Sciences EZBio 250mL PETG Media Bottles, Sterile, Square Storage Bottles with 38-430mm White Closed VersaCap Closures, 24/PK
Catalog No. 501613226
Encompass_Preferred
EZBio PETG Sterile Media Bottles are perfect for storage of sterile reagents and media, and can be used with EZBio Single-Use Cap Assemblies for aseptic transfer and sampling applications. Bottles withstand temperatures down to -40C for freezing applications. Gamma-irradiated. Assembled in a Class 10,000 Cleanroom. Packaged in double-bagged trays. 100% USP Class VI, BPA-free, ADCF-free materials. VersaCaps with vented membranes or molded hose barbs available separately.
Catalog No. 50-161-3226 Supplier Foxx Life Sciences Supplier No. 1373211FLS
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
EZBio PETG Sterile Media Bottles are perfect for storage of sterile reagents and media, and can be used with EZBio Single-Use Cap Assemblies for aseptic transfer and sampling applications. Bottles withstand temperatures down to -40C for freezing applications. Gamma-irradiated. Assembled in a Class 10,000 Cleanroom. Packaged in double-bagged trays. 100% USP Class VI, BPA-free, ADCF-free materials. VersaCaps with vented membranes or molded hose barbs available separately.