Learn More
Research Products International Corp PETG Media Bottle, 1000ml, 24/cs
SDP
Catalog No. 501977449
Encompass
These square PETG media bottles offer the clarity of glass with the durability of plastic and a space saving design. Improved features include a wide mouth opening, thicker wall dimensions and a leak proof double seal closure. Bottles have permanent molded in graduations and dripless pour lip. Bottles and caps are pre-sterilized and tested for pH stability, temperature durability, cloning efficiency and cytotoxicity. Supplied with caps attached in shrink-wrapped trays.
Catalog No. 50-197-7449 Supplier Research Products International Corp Supplier No. 219980
These square PETG media bottles offer the clarity of glass with the durability of plastic and a space saving design. Improved features include a wide mouth opening, thicker wall dimensions and a leak proof double seal closure. Bottles have permanent molded in graduations and dripless pour lip. Bottles and caps are pre-sterilized and tested for pH stability, temperature durability, cloning efficiency and cytotoxicity. Supplied with caps attached in shrink-wrapped trays.