Learn More
Alkali Scientific PETG Media Bottles Square with
SDP
Supplier:  Alkali Scientific VB250PETG
Encompass_Preferred
Our Square Reagent Bottle are an ideal option for storage and transportation container for culture medium serum buffer intermediates and reagents for biopharmaceutical and biotechnology companies reagent RandD institutions etc The raw materials of the bottle PETG are in line with USP Class VI Class USP661 biocompatibility requirements and ISO10993 Quality System Product Features1 PET wall is light in weight and resistant to low temperature while PETG wall is thick durable and resistant to fracture 2 High chemical durability suitable for the storage of high purity reagent standard particle materials and samples 3 High transparency strong mechanical strength and shock resistance and convenient for observation and transportation4 Highly tolerant of low temperature UV rays and resistant to fracture5 Smooth inner surface can minimize the residue on the wall6 Sterile DNase/RNase/Pyrogen free MaterialPETG Bottle and HDPE Cap ApplicationIt is mainly used for storing and sampli
Catalog No. 50-302-8155