Learn More
VWR INTERNATIONAL LLC PETG SINGLE-USE BOTTLE ASSEMBL
Catalog No. NC2232658
Encompass
1000 ml (32 oz.) Port 1: PP Tube Plug Port 2: 25mm Hydrophobic PE Vent Filter C-Flex Port 1: 24" Port 2: 3" 38-430 Port 1: 1/8 x 1/4" Port 2: 1/8 x 1/4"
This item is not returnable. View return policy
Catalog No. NC2232658 Supplier VWR INTERNATIONAL LLC Supplier No. 10830298
1000 ml (32 oz.) Port 1: PP Tube Plug Port 2: 25mm Hydrophobic PE Vent Filter C-Flex Port 1: 24" Port 2: 3" 38-430 Port 1: 1/8 x 1/4" Port 2: 1/8 x 1/4"